Order Entry
Export
Orders LinkContactUsLinkComponent
Estradiol 98%

Estradiol 98%

Glass bottle, 5 g
Catalog # ACRO436320050
Health
Estradiol 98%
Catalog # ACRO436320050
CAS Number:  50-28-2
Estradiol 98%
Estradiol 98%

Glass bottle, 5 g

CAS Number:  50-28-2
Price
172,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • MDL Number:MFCD00003693
  • Molecular Weight:272.39 g/mol
  • Melting Pt:176…180 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    173 to 179 °C
  • Water:
    ≤3.5 %
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +76° to +83° (20 °C, 589 nm) (c=1, dioxane)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications