Order Entry
Export
Orders LinkContactUsLinkComponent
2,2-Dimethylcyclohexanone

2,2-Dimethylcyclohexanone

Glass bottle, 5 g
Catalog # ACRO426500050
Irritant
Flammable
2,2-Dimethylcyclohexanone
Catalog # ACRO426500050
CAS Number:  1193-47-1
2,2-Dimethylcyclohexanone
2,2-Dimethylcyclohexanone

Glass bottle, 5 g

CAS Number:  1193-47-1
Price
864,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1193-47-1
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00134382
  • Molecular Weight:126.1962 g/mol
  • Boiling Pt:169…170 °C (.8 mmHg)
  • Melting Pt:–20 °C
  • Density:0.912 g/cm³
  • Flash Pt:50 °C
  • Storage Temperature:Ambient
  • UN:1224
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4470 to 1.4490 (20°C, 589 nm)
  • GC:
    ≥94 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications