2,2-Dimethylcyclohexanone
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
2,2-Dimethylcyclohexanone
Catalog # ACRO426500010
Supplier: Thermo Fisher Scientific
CAS Number: 1193-47-1
Jump to:
Product Details & Documents
- CAS Number:1193-47-1
- Formula:C₈H₁₄O
- MDL Number:MFCD00134382
- Molecular Weight:126.1962 g/mol
- Boiling Pt:169…170 °C (.8 mmHg)
- Melting Pt:–20 °C
- Density:0.912 g/cm³
- Flash Pt:50 °C
- Storage Temperature:Ambient
- UN:1224
- ADR:3,III
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4470 to 1.4490 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



