Sodium nitrate, extra pure
Fibre drum, 25 kg
Supplier: Thermo Fisher Scientific
Sodium nitrate, extra pure
Catalog # ACRO436090250
Supplier: Thermo Fisher Scientific
CAS Number: 7631-99-4
Jump to:
Product Details & Documents
- CAS Number:7631-99-4
- Formula:NaNO₃
- MDL Number:MFCD00011119
- Molecular Weight:84.99 g/mol
- Boiling Pt:380 °C (760 mmHg)
- Melting Pt:306 °C
- Density:2,26 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:1498
- ADR:5.1,III
Specifications
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Loss on drying:≤2 % (105°C)
- pH:5.5 to 8.3 (5 % in water)
- Titration after Ion exchange:≥99.0 %
Specifications
- Pack type:Fibre drum
- Pk:25 kg



