3,5-Dihydroxybenzaldehyde 98%
Glass bottle, 5 g
- :26153-38-8
- :C₇H₆O₃
- :MFCD00016611
- :138.12 g/mol
- :153…158 °C
- :Ambient
- Appearance:White to grey to tan Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 %
- HPLC:≥97.5 %
- Pack type:Glass bottle
- Pk:5 g



