3,5-Dihydroxybenzaldehyde 98%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
3,5-Dihydroxybenzaldehyde 98%
Catalog # ACRO425650010
Supplier: Thermo Fisher Scientific
CAS Number: 26153-38-8
Jump to:
Product Details & Documents
- CAS Number:26153-38-8
- Formula:C₇H₆O₃
- MDL Number:MFCD00016611
- Molecular Weight:138.12 g/mol
- Melting Pt:153…158 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to grey to tan Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 %
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



