Order Entry
Export
Orders LinkContactUsLinkComponent
Anthrone 95%, pure

Anthrone 95%, pure

Glass bottle, 100 g
Catalog # ACRO401361000
Irritant
Anthrone 95%, pure
Catalog # ACRO401361000
CAS Number:  90-44-8
Anthrone 95%, pure
Anthrone 95%, pure

Glass bottle, 100 g

CAS Number:  90-44-8
Price
219,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Boiling Pt:721 °C (760 mmHg)
  • Melting Pt:152…157 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to beige Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    155 to 161 °C
  • GC:
    ≥94 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications