Methyl oleate, technical
Plastic drum, 5 kg
: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
- :67762-38-3
- :C₁₉H₃₆O₂
- :MFCD00009578
- :296.49 g/mol
- :12,06965
- :186 °C (9 mmHg)
- :–20 °C
- :0.87 g/cm³
- :170 °C
- :Refrigerator
- :203-992-5
- Appearance:Clear yellow to amber Viscous liquid
- Infrared spectrum:Conforms
- GC:≥55 %
- HPLC:≥98.0 %
- Pack type:Plastic drum
- Pk:5 kg



