MBD 98%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 33984-50-8
Synonyms: 7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole
- CAS Number:33984-50-8
- Formula:C₁₄H₁₂N₄O₄
- MDL Number:MFCD00042778
- Molecular Weight:300.274 g/mol
- Melting Pt:176…180 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:Orange to brown Powder
- Infrared spectrum:Conforms
- Melting point:177 to 182 °C
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



