3-(4-Bromophenyl)propionic acid 97%
Glass bottle, 5 g
- :1643-30-7
- :C₉H₉BrO₂
- :MFCD01310793
- :229.07 g/mol
- :133…136 °C
- :Ambient
- :3335
- :9,III
- Appearance:White to yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:134 to 138 °C
- HPLC:≥96.0 %
- Pack type:Glass bottle
- Pk:5 g



