3-(4-Bromophenyl)propionic acid 97%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
3-(4-Bromophenyl)propionic acid 97%
Catalog # ACRO399450010
Supplier: Thermo Fisher Scientific
CAS Number: 1643-30-7
Jump to:
Product Details & Documents
- CAS Number:1643-30-7
- Formula:C₉H₉BrO₂
- MDL Number:MFCD01310793
- Molecular Weight:229.07 g/mol
- Melting Pt:133…136 °C
- Storage Temperature:Ambient
- UN:3335
- ADR:9,III
Specifications
Specification Test Results
- Appearance:White to yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:134 to 138 °C
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



