1,3-Diiodobenzene 98%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
1,3-Diiodobenzene 98%
Catalog # ACRO407740250
Supplier: Thermo Fisher Scientific
CAS Number: 626-00-6
Jump to:
Product Details & Documents
- CAS Number:626-00-6
- Formula:C₆H₄I₂
- MDL Number:MFCD00041731
- Molecular Weight:329.91 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:33…37 °C
- Density:2.47 g/cm³
- Storage Temperature:Refrigerator
- EINECS:210-921-1
Documents
ACRO40774
SDS
Specifications
Specification Test Results
- Appearance:Yellow to beige Powder or crystals or low melting crystalline mass
- Infrared spectrum:Conforms
- Melting point:33 to 37 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



