Cellulose, microcrystalline ∼ Ø 50 µm
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Cellulose, microcrystalline ∼ Ø 50 µm
Catalog # ACRO387240050
Supplier: Thermo Fisher Scientific
CAS Number: 9004-34-6
Jump to:
Product Details & Documents
Synonyms: Microcrystallinecellulose
- CAS Number:9004-34-6
- Formula:(C₁₂H₂₂O₁₁)n
- MDL Number:MFCD00081512
- Molecular Weight:342.3 g/mol
- Merck Index:13,01977
- Melting Pt:260 - 270 °C
- Density:1,27 to 1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-674-9
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1.5 %
- Particle size:≤1 % (> 250 µm)
- Residue after ignition:≤0.05 %
- pH:5 to 7
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic drum
- Pk:5 kg
- Cat. No.:ACRO387240050



