Formamide 99+% for spectroscopy
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Formamide 99+% for spectroscopy
Catalog # ACRO410760025
Supplier: Thermo Fisher Scientific
CAS Number: 75-12-7
Jump to:
Product Details & Documents
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (760 mmHg)
- Melting Pt:2…3 °C
- Density:1.133 g/cm³
- Flash Pt:175 °C
- Storage Temperature:Ambient
- EINECS:200-842-0
Documents
ACRO41076
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 % (Coulometric)
- GC:≥99.0 %
- UV:at 275 nm A: ≤0.5
- Color scale:≤10 APHA
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



