Formamide 99+% for spectroscopy
Glass bottle, 1 L
: Carbamaldehyde
- :75-12-7
- :HCONH₂
- :MFCD00007941
- :45.04 g/mol
- :13,04261
- :210 °C (760 mmHg)
- :2…3 °C
- :1.133 g/cm³
- :175 °C
- :Ambient
- :200-842-0
- Appearance:Clear liquid
- Water:≤0.03 % (Coulometric)
- GC:≥99.0 %
- UV:at 275 nm A: ≤0.5
- Color scale:≤10 APHA
- Pack type:Glass bottle
- Pk:1 L



