Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl ether ≥99% stabilised for HPLC

Diethyl ether ≥99% stabilised for HPLC

Glass bottle, 2,5 L
Catalog # ACRO410040025
Irritant
Flammable
Diethyl ether ≥99% stabilised for HPLC
Catalog # ACRO410040025
CAS Number:  60-29-7
Diethyl ether ≥99% stabilised for HPLC
Diethyl ether ≥99% stabilised for HPLC

Glass bottle, 2,5 L

CAS Number:  60-29-7
Price
148,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–116 °C
  • Density:0.714 g/cm³
  • Flash Pt:–45 °C
  • Storage Temperature:Ambient
  • UN:1155
  • ADR:3,I
  •  :Stabilised with ethanol

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 %
  • GC:
    ≥99 %
  • UV:
    at 210 nm A: ≤0.3
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    2 % Ethanol
  • Wavelength at 1 A.U.:
    ≤205 nm
  • Residue after evaporation:
    ≤2 ppm
  • Acidity/alkalinity:
    ≤0.0002 meq/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications