Glass Wool
Glass wool
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
This glass wool can be used as a filter aid.
Boiling point: >1000 °C (760 mmHg)
Documents
ACRO38606
SDS
Specifications
- Description:Glass wool
- Cat. No.:ACRO386062500