Orders LinkContactUsLinkComponent
Glass Wool
Glass Wool
Plastic drum, Glass wool
Catalog # ACRO386060010
Glass Wool
Catalog # ACRO386060010
Glass Wool
Glass Wool
Plastic drum, Glass wool
Price
97,40 €
Log in for availability
1 * 1 kg
Product Details & Documents
This glass wool can be used as a filter aid.
Boiling point: >1000 °C (760 mmHg)
Documents
Specifications
  • Description:
    Glass wool
  • Pack type:
    Plastic drum