2-Nitroethanol 97%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:625-48-9
- Formula:C₂H₅NO₃
- MDL Number:MFCD00007405
- Molecular Weight:91.07 g/mol
- Boiling Pt:203…206 °C (760 mmHg)
- Melting Pt:–80 °C
- Density:1.292 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
Documents
ACRO39796
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4430 to 1.4450 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 g



