4-Iodopyridine 96%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:15854-87-2
- Formula:C₅H₄IN
- MDL Number:MFCD02093937
- Molecular Weight:205 g/mol
- Melting Pt:92…96 °C
- Storage Temperature:Freezer
Documents
ACRO39129
SDS
Specifications
Specification Test Results
- Appearance:Light brown to dark green-brown Crystals or powder (May darken on storage)
- Infrared spectrum:Conforms
- GC:≥95 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



