4,4'-Biphenyldicarboxaldehyde 97%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
4,4'-Biphenyldicarboxaldehyde 97%
Catalog # ACRO391130050
Supplier: Thermo Fisher Scientific
CAS Number: 66-98-8
Jump to:
Product Details & Documents
Synonyms: [1,1'-Biphenyl]-4,4'-dicarbaldehyde, 4,4'-Diformylbiphenyl
- CAS Number:66-98-8
- Formula:C₁₄H₁₀O₂
- MDL Number:MFCD00016714
- Molecular Weight:210.23 g/mol
- Melting Pt:144…147 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:144 to 147 °C
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



