ω-Bromo-4-fluoroacetophenone 98%
Glass bottle, 25 g
: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
- :403-29-2
- :C₈H₆BrFO
- :MFCD00040830
- :217.04 g/mol
- :150 °C (12 mmHg)
- :46…49 °C
- :>110 °C
- :Refrigerator
- :206-955-1
- :3261
- :8,III
- Appearance:White to yellow to yellow-green to beige Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:45 to 50 °C
- GC:≥96.0 %
- Pack type:Glass bottle
- Pk:25 g



