2,3-Dimethylhydroquinone 97%
Glass bottle, 100 g
: 1,4-Dihydroxy-2,3-dimethylbenzene
- :608-43-5
- :C₈H₁₀O₂
- :MFCD00009997
- :138.17 g/mol
- :223…225 °C
- :Ambient
Documents
ACRO30511
SDS
- Appearance:Off-white to light brown to purple Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Pack type:Glass bottle
- Pk:100 g



