2,3-Dimethylhydroquinone 97%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
2,3-Dimethylhydroquinone 97%
Catalog # ACRO305110250
Supplier: Thermo Fisher Scientific
CAS Number: 608-43-5
Jump to:
Product Details & Documents
Synonyms: 1,4-Dihydroxy-2,3-dimethylbenzene
- CAS Number:608-43-5
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00009997
- Molecular Weight:138.17 g/mol
- Melting Pt:223…225 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Off-white to light brown to purple Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



