Acetonitrile 99.5% for analysis
Metal drum, 25 L
Supplier: Thermo Fisher Scientific
Acetonitrile 99.5% for analysis
Catalog # ACRO258560250
Supplier: Thermo Fisher Scientific
CAS Number: 75-05-8
Jump to:
Product Details & Documents
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS Number:75-05-8
- Formula:H₃CCN
- MDL Number:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Boiling Pt:81…82 °C (760 mmHg)
- Melting Pt:–46 °C
- Density:0.781 g/cm³
- Flash Pt:2 °C
- Storage Temperature:Ambient
- EINECS:200-835-2
- UN:1648
- ADR:3,II
- REACH:01-2119471307-38
Documents
ACRO25856
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 % (Coulometric)
- GC:≥99.4 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.005 %
- Calcium (Ca):≤1 ppm
- Copper (Cu):≤2 ppm
- Iron (Fe):≤2 ppm
- Lead (Pb):≤2 ppm
- Magnesium (Mg):≤0.5 ppm
- Potassium (K):≤1 ppm
- Sodium (Na):≤5 ppm
- Zinc (Zn):≤2 ppm
- Free acid (CH3COOH):≤0.005 %
- Free alkali (NH3):≤0.001 %
Specifications
- Pack type:Metal drum
- Pk:25 L
- Cat. No.:ACRO258560250
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health



