4-Hydroxy-2-methoxybenzaldehyde 98%
Glass bottle, 25 g
- :18278-34-7
- :C₈H₈O₃
- :MFCD00051964
- :152.15 g/mol
- :273 °C (1013 hPa)
- :153…158 °C
- :Ambient
- Appearance:Off-white to light brown Powder
- Infrared spectrum:Conforms
- Melting point:156 to 161 °C
- HPLC:≥97.5 %
- Pack type:Glass bottle
- Pk:25 g



