Order Entry
Export
Orders LinkContactUsLinkComponent
Behenic acid 85%, Technical Grade

Behenic acid 85%, Technical Grade

Glass bottle, 250 g
Catalog # ACRO334812500
Behenic acid 85%, Technical Grade
Catalog # ACRO334812500
CAS Number:  112-85-6
Behenic acid 85%, Technical Grade
Behenic acid 85%, Technical Grade

Glass bottle, 250 g

CAS Number:  112-85-6
Price
50,20 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:13,01024
  • Boiling Pt:306 °C (60 mmHg)
  • Melting Pt:80…82 °C
  • Density:0,8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥80 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g

Specifications