D-(+)-Maltose monohydrate 92+%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
D-(+)-Maltose monohydrate 92+%
Catalog # ACRO329910025
Supplier: Thermo Fisher Scientific
CAS Number: 6363-53-7
Jump to:
Product Details & Documents
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
- CAS Number:6363-53-7
- Formula:C₁₂H₂₂O₁₁·1H₂O
- MDL Number:MFCD00149343
- Molecular Weight:360.32 g/mol
- Merck Index:13,05736
- Melting Pt:130 °C
- Storage Temperature:Ambient
- EINECS:200-716-5
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤7 %
- Sulfated ash:≤0.1 %
- Additional info:remainder mainly glucose and maltotriose
- HPLC:≥92.0 %
- Specific optical rotation:+135° to +139° (20°C, 589 nm) (c=4, H2O, NH3) (on dry substance)
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



