n-Heptane, extra dry 99+%, AcroSeal®
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
n-Heptane, extra dry 99+%, AcroSeal®
Catalog # 326910010.
Supplier: Thermo Fisher Scientific
CAS Number: 142-82-5
Jump to:
Product Details & Documents
Synonyms: high purity solvents
- CAS Number:142-82-5
- Formula:H₃C(CH₂)₅CH₃
- MDL Number:MFCD00009544
- Molecular Weight:100.20 g/mol
- Merck Index:13,04679
- Boiling Pt:98 °C (760 mmHg)
- Melting Pt:–91 °C
- Density:0.683 g/cm³
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-563-8
- UN:1206
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.0005 %
Specifications
- Pack type:Glass bottle
- Pk:1 L



