Cadmium acetate dihydrate 98% for analysis
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Cadmium acetate dihydrate 98% for analysis
Catalog # ACRO317130025
Supplier: Thermo Fisher Scientific
CAS Number: 5743-04-4
Jump to:
Product Details & Documents
Synonyms: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate
- CAS Number:5743-04-4
- Formula:(H₃CCOO)₂Cd·2H₂O
- MDL Number:MFCD00150020
- Molecular Weight:266.53 g/mol
- Merck Index:13,01614
- Storage Temperature:Ambient
- EINECS:208-853-2
- UN:2570
- ADR:6.1,III
Documents
ACRO31713
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Complexometric:≥97.5 %
- Chloride (Cl):≤10 ppm
- Nitrate (NO3):≤30 ppm
- Sulfate (SO4):≤50 ppm
- Copper (Cu):≤10 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤50 ppm
- Zinc (Zn):≤50 ppm
- Insoluble matter:≤0.005 % (in water)
- Subst not pptd by H2S:≤0.3 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



