2,7-Dibromofluorene 99%
Glass bottle, 25 g
: 2,7-Dibromo-9H-fluorene
- :16433-88-8
- :C₁₃H₈Br₂
- :MFCD00019048
- :324.01 g/mol
- :164…167 °C
- :Ambient
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:164 to 167 °C
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:25 g



