Hydrocortisone 98%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:50-23-7
- Formula:C₂₁H₃₀O₅
- MDL Number:MFCD00011654
- Molecular Weight:362.47 g/mol
- Boiling Pt:567 °C (1013 hPa)
- Melting Pt:212…217 °C
- Density:1,4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-020-1
Specifications
Specification Test Results
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Melting point:212 to 217 °C
- HPLC:≥97.5 %
- Specific optical rotation:+150° to +156° (20 °C, 589 nm) (C=1, dioxane)
Specifications
- Pack type:Glass bottle
- Pk:25 g



