3,4-Dichlorophenethylamine 99%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
3,4-Dichlorophenethylamine 99%
Catalog # ACRO350800250
Supplier: Thermo Fisher Scientific
CAS Number: 21581-45-3
Jump to:
Product Details & Documents
- CAS Number:21581-45-3
- Formula:C₈H₉Cl₂N
- MDL Number:MFCD00060617
- Molecular Weight:190.07 g/mol
- Boiling Pt:280 °C
- Density:1.268 g/cm³
- Flash Pt:110 °C
- Storage Temperature:Ambient
- ADR:8,III
Documents
ACRO35080
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5655 to 1.5675 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



