Uric acid 99+%
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione
- CAS Number:69-93-2
- Formula:C₅H₄N₄O₃
- MDL Number:MFCD00005712
- Molecular Weight:168.11 g/mol
- Merck Index:12,10014
- Melting Pt:>300 °C
- Density:1,89 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-720-7
Documents
ACRO17129
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Assay:≥99.0 % (ex N)
- Loss on drying:≤0.5 % (105°C, 3 hrs)
- Sulfated ash:≤0.2 %
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



