Nalidixic acid 99.5%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:389-08-2
- Formula:C₁₂H₁₂N₂O₃
- MDL Number:MFCD00006884
- Molecular Weight:232.24 g/mol
- Melting Pt:225…231 °C
- Storage Temperature:Ambient
- EINECS:206-864-7
Documents
ACRO16990
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:225 to 231 °C
- Titration with NaOH:≥99.4 %
- Specific optical rotation:-44.0° to -48.0° (20 °C, 589 nm) (c=0.7, water)
Specifications
- Pack type:Glass bottle
- Pk:100 g



