Bis-trispropane 99%
Glass bottle, 250 g
Supplier: Thermo Fisher Scientific
Bis-trispropane 99%
Catalog # ACRO202642500
Supplier: Thermo Fisher Scientific
CAS Number: 64431-96-5
Jump to:
Product Details & Documents
Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
- CAS Number:64431-96-5
- Formula:C₁₁H₂₆N₂O₆
- MDL Number:MFCD00004689
- Molecular Weight:282.34 g/mol
- Melting Pt:164…165 °C
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:161 to 168 °C
- Loss on drying:≤1 %
- Solubility:(0.1 M in water) Clear colorless
- UV:(0.1 M in water)
- Titration with HClO4:98.5 to 101.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 g



