Order Entry
Export
Orders LinkContactUsLinkComponent
Adenosine 99+%

Adenosine 99+%

Glass bottle, 500 g
Catalog # ACRO164045000
Adenosine 99+%
Catalog # ACRO164045000
CAS Number:  58-61-7
Adenosine 99+%
Adenosine 99+%

Glass bottle, 500 g

CAS Number:  58-61-7
Price
1 100,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Melting Pt:233…238 °C
  • Storage Temperature:Ambient
  • EINECS:200-389-9

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    233 to 238 °C
  • Loss on drying:
    ≤0.5 % (105 °C, 2 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥99.0 %
  • UV spectrophotometry:
    98.0 to 102.0 % (lambda max. 259 nm, pH 7)
  • UV transmittance:
    ≥98 % (c=0.5, H2O, 430 nm) 1 cm cell
  • Ratio of Absorbances:
    0.76 to 0.80 (A250/A260)
  • Nitrogen (N):
    25.69 to 26.73 % (on dry basis)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications