1,3-Dithiane 97%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:505-23-7
- Formula:C₄H₈S₂
- MDL Number:MFCD00006654
- Molecular Weight:120.24 g/mol
- Boiling Pt:207 - 208 °C (1013 hPa)
- Melting Pt:49…54 °C
- Density:1,36 g/cm³ (25 °C)
- Flash Pt:90 °C
- Storage Temperature:Ambient
- EINECS:208-006-7
- ADR:9,
- :Density: 1.36 g/cm³ (25 °C)
Documents
ACRO16951
SDS
Specifications
Specification Test Results
- Appearance:White to light beige Powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:49 to 54 °C
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 g



