1,3-Dihydroxynaphthalene 99+%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
1,3-Dihydroxynaphthalene 99+%
Catalog # ACRO163340250
Supplier: Thermo Fisher Scientific
CAS Number: 132-86-5
Jump to:
Product Details & Documents
Synonyms: Naphthalene-1,3-diol, Naphthalene-1,3-diol
- CAS Number:132-86-5
- Formula:C₁₀H₈O₂
- MDL Number:MFCD00003965
- Molecular Weight:160.17 g/mol
- Merck Index:12,06483
- Boiling Pt:386 °C (1013 hPa)
- Melting Pt:123…126 °C
- Density:1,12 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-079-7
Documents
Specifications
Specification Test Results
- Appearance:Pink-orange to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:123 to 126 °C
- Titration with NaOH:≥99.0 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



