Lead(II) oxide 99.7%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Lead(II) oxide 99.7%
Catalog # ACRO199340025
Supplier: Thermo Fisher Scientific
CAS Number: 1317-36-8
Jump to:
Product Details & Documents
Synonyms: Lead monoxide
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:13,05431
- Boiling Pt:1470 °C
- Melting Pt:886 °C
- Density:9,5 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-267-0
- UN:3077
- ADR:9,III
Documents
ACRO19934
SDS
Specifications
Specification Test Results
- Appearance:Yellow Powder
- Titration Complexometric:≥99.6 %
- Loss on drying:≤0.1 % (105°C)
- Copper (Cu):≤10 ppm
- Iron (Fe):≤20 ppm
- Lead (Pb):≤0.01 %
- Silicon (Si):≤20 ppm
- Lead oxide (Pb3O4):≤0.02 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



