Lead(IV) oxide 97+%, extra pure
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Lead(IV) oxide 97+%, extra pure
Catalog # ACRO217530025
Supplier: Thermo Fisher Scientific
CAS Number: 1309-60-0
Jump to:
Product Details & Documents
Synonyms: Lead dioxide
- CAS Number:1309-60-0
- Formula:O₂Pb
- MDL Number:MFCD00011165
- Molecular Weight:239.2 g/mol
- Merck Index:12,05424
- Melting Pt:290 °C (dec.)
- Density:9,4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-174-5
- UN:1872
- ADR:5.1,III
Documents
Specifications
Specification Test Results
- Appearance:Brown to black Powder
- Titration with Na2S2O3:≥97.0 %
- Chloride (Cl):≤0.02 %
- Sulfate (SO4):≤0.005 %
- Copper (Cu):≤0.01 %
- Iron (Fe):≤0.01 %
- Zinc (Zn):≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



