2,5-Dichloropyridine 98%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
2,5-Dichloropyridine 98%
Catalog # ACRO186141000
Supplier: Thermo Fisher Scientific
CAS Number: 16110-09-1
Jump to:
Product Details & Documents
- CAS Number:16110-09-1
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00006239
- Molecular Weight:147.99 g/mol
- Boiling Pt:190…191 °C (760 mmHg)
- Melting Pt:58…62 °C
- Flash Pt:112 °C
- Storage Temperature:Ambient
- EINECS:240-278-2
- ADR:6.1,III
Documents
ACRO18614
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:58 to 62 °C
- Water:≤1 %
- GC:≥97.5 %
- Titration with NaOH:28.1 to 29.5 % (citric acid dry substance)
Specifications
- Pack type:Glass bottle
- Pk:100 g



