Order Entry
Export
Orders LinkContactUsLinkComponent
n-Hexane 95+% for HPLC

n-Hexane 95+% for HPLC

Glass bottle, 2,5 L
Catalog # ACRO232100025
Irritant
Health
Environment
Flammable
n-Hexane 95+% for HPLC
Catalog # ACRO232100025
CAS Number:  110-54-3
n-Hexane 95+% for HPLC
n-Hexane 95+% for HPLC

Glass bottle, 2,5 L

CAS Number:  110-54-3
Price
206,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Boiling Pt:69 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.659 g/cm³
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • UN:1208
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.01 %
  • GC:
    ≥95.0 %
  • UV:
    at 200 nm A: ≤0.7
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.0002 meq/g
  • Filtered to 0.2 micrometer:
    Filtered

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications