Potassium cyanide 97+% for analysis
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Potassium cyanide 97+% for analysis
Catalog # ACRO196601000
Supplier: Thermo Fisher Scientific
CAS Number: 151-50-8
Jump to:
Product Details & Documents
- CAS Number:151-50-8
- Formula:KCN
- MDL Number:MFCD00011397
- Molecular Weight:65.12 g/mol
- Merck Index:13,07709
- Boiling Pt:1625 °C
- Melting Pt:634 °C
- Density:1,55 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-792-3
- UN:1680
- ADR:6.1,I
Documents
ACRO19660
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥97.0 %
- Chloride (Cl):≤500 ppm
- Thiocyanate (SCN):≤100 ppm
- Phosphate (PO4):≤50 ppm
- Sulfate (SO4):≤200 ppm
- Iron (Fe):≤100 ppm
- Lead (Pb):≤5 ppm
- Sodium (Na):≤5000 ppm
- Sulfur (S):≤10 ppm
Specifications
- Pack type:Plastic bottle
- Pk:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



