Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thymidine (2'-deoxythymidine) 99+%

Glass bottle, 25 g
Catalog # ACRO226740250
Health
Irritant
Thymidine (2'-deoxythymidine) 99+%
Catalog # ACRO226740250
CAS Number:  50-89-5
Thymidine (2'-deoxythymidine) 99+%
Thymidine (2'-deoxythymidine) 99+%

Glass bottle, 25 g

CAS Number:  50-89-5
Price
583,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • CAS Number:50-89-5
  • Formula:C₁₀H₁₄N₂O₅
  • MDL Number:MFCD00006537
  • Molecular Weight:242.23 g/mol
  • Melting Pt:187…189 °C
  • Storage Temperature:Ambient
  • EINECS:200-070-4

 

Specifications

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 % (K.F.)
  • Sulfated ash:
    ≤0.3 % (800°C)
  • HPLC:
    ≥99.0 %
  • UV spectrophotometry:
    98.0 to 102.0 % (lambda max. 267 nm, pH 7)
  • Ratio of Absorbances:
    0.62 to 0.68 (A250/A260)
  • Specific optical rotation:
    +17.5° to +19.5° (20°C, 589 nm) (c=3, H2O)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications