Zinc oxide 99.5+%
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Zinci oxidum, Zinc (II) oxide
- CAS Number:1314-13-2
- Formula:ZnO
- MDL Number:MFCD00011300
- Molecular Weight:81.39 g/mol
- Merck Index:13,10200
- Melting Pt:1975 °C
- Density:5.6 g/cm³
- Storage Temperature:Ambient
- EINECS:215-222-5
- UN:3077
- ADR:9,III
- REACH:01-2119463881-32
Documents
ACRO19489
SDS
Specifications
Specification Test Results
- Appearance:White to faintly yellowish-white Powder
- Titration Complexometric:≥99.50 %
- Additional info:Carbonates and substances insoluble in acids:
- Particle size:≤0.01 % Sieve residue on 45 micron sieve
- Residue after ignition:≤1 % (1 g, 500°C)
- Specific surface area:3.5 to 9 m²/g
- Alkalinity:≤0.3 ml/g (ml 0.1 N HCl/g)
- Arsenic (As):≤5 ppm
- Cadmium (Cd):≤10 ppm
- Iron (Fe):≤200 ppm
- Lead (Pb):≤50 ppm
Specifications
- Pack type:Plastic drum
- Pk:5 kg



