Sodium alginate
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Alginic acid sodium salt, Algin
- CAS Number:9005-38-3
- MDL Number:MFCD00081310
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:Off-white to yellow-brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15.0 % (100 mg, 105°C, 4 hrs)
- Sulfated ash:30.0 % to 36.0 % (on dried substance)
- Viscosity:350 to 550 mPa.s (1 % at 20°C) (Brookfield LV)
- pH:5.5 to 8.0 (1 % at 20°C)
- Heavy metals (as Pb):≤20 ppm
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



