Lead(II) carbonate ACS
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Lead(II) carbonate ACS
Catalog # ACRO212515000
Supplier: Thermo Fisher Scientific
CAS Number: 598-63-0
Jump to:
Product Details & Documents
Synonyms: Lead carbonat
- CAS Number:598-63-0
- Formula:PbCO₃
- MDL Number:MFCD00064631
- Molecular Weight:267.21 g/mol
- Melting Pt:400 °C
- Density:6,43 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:White Powder
- Chloride (Cl):≤0.002 %
- Nitrate and Nitrite (as NO3):(limit about 0.005%) Passes test
- Cadmium (Cd):≤0.002 %
- Calcium (Ca):≤0.01 %
- Iron (Fe):≤0.005 %
- Potassium (K):≤0.02 %
- Sodium (Na):≤0.05 %
- Zinc (Zn):≤0.003 %
- Insoluble matter:≤0.02 % (in dilute acetic acid)
Specifications
- Pack type:Plastic bottle
- Pk:500 g



