Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) chloride 99%

Lead(II) chloride 99%

Plastic bottle, 50 g
Catalog # ACRO193310500
Irritant
Health
Environment
Lead(II) chloride 99%
Catalog # ACRO193310500
CAS Number:  7758-95-4
Lead(II) chloride 99%
Lead(II) chloride 99%

Plastic bottle, 50 g

CAS Number:  7758-95-4
Price
20,20 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Lead dichloride
  • CAS Number:7758-95-4
  • Formula:Cl₂Pb
  • MDL Number:MFCD00011157
  • Molecular Weight:278.11 g/mol
  • Boiling Pt:950 °C (1013 hPa)
  • Melting Pt:501 °C
  • Density:5,85 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:2291
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Titration Complexometric:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g

Specifications