Order Entry
Export
Orders LinkContactUsLinkComponent
 

Progesterone 98%

Glass bottle, 100 g
  ACRO225651000
 
Progesterone 98%
  ACRO225651000
 :  57-83-0
Progesterone 98%
Progesterone 98%

Glass bottle, 100 g

 :  57-83-0
 
339,00 €
   
1 * 100 g
 
  •  :57-83-0
  •  :C₂₁H₃₀O₂
  •  :MFCD00003658
  •  :126…131 °C
  •  :Ambient
  •  :200-350-6
 

 

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    126 to 131 °C
  • Loss on drying:
    ≤0.5 % (500 mg, 105 °C, 2 hrs)
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +186° to +194° (20 °C, 589 nm) (c=1, ethanol) (on dried substance)

 

  • Pack type:
    Glass bottle
  • Pk:
    100 g